missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.0100 N (N/100), Ricca Chemical

Supplier:  Ricca Chemical Company 619632

 View more versions of this product

Catalog No. 6196-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
1310-58-3,7732-18-5
100.0,0.07
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
Laboratory
Liquid
Colorless
98.0,0.06
HKO
0.0100Normal
1 L
[OH-].[K+]
56.11
CHEBI:32035
Natural Poly Bottle

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions