missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.0100 N (N/100) in Methanol, Ricca Chemical
$140.56 - $543.59
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| IUPAC Name | potassium hydroxide |
| SMILES | [OH-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R6264000500
|
Ricca Chemical Company
R6264000500A |
500 mL | Natural Poly Bottle |
Each for $140.56
|
|
||||
|
R62640001A
|
Ricca Chemical Company
R62640001A |
1 L | Natural Poly Bottle |
Each for $522.91
|
|
||||
|
R62640004A
|
Ricca Chemical Company
R62640004A |
4 L | Natural Poly Bottle |
Each for $543.59
|
|
||||
Chemical Identifiers
| 67-56-1 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide |
Specifications
| Colorless | |
| 0.01000 ± 0.00005 N at 20°C | |
| MFCD00003553 | |
| Solubility in Water: Infinite | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| Laboratory | |
| Natural Poly Bottle | |
| Liquid |
| 67-56-1,1310-58-3 | |
| HKO | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| 500 mL | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Characteristic alcohol | |
| Nonaqueous Solution | |
| Potassium Hydroxide, 0.0100 Normal (N/100) in Methanol |
Safety and Handling
ShelfLife : 12 months
Recommended Storage : Ambient Flammable