missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroquinone Monosulfate 98.0+%, TCI America™

Catalog No. H018710G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H018710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H018710G Supplier TCI America Supplier No. H018710G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37067-27-9
Molecular Formula C6H5KO5S
Molecular Weight (g/mol) 228.259
MDL Number MFCD00059841
InChI Key KYQHUXZXYUBUPX-UHFFFAOYSA-M
Synonym Hydroquinone Monosulfate Potassium Salt
PubChem CID 101973306
IUPAC Name potassium;4-sulfooxyphenolate
SMILES C1=CC(=CC=C1[O-])OS(=O)(=O)O.[K+]

Specifications

Quantity 10 g
Formula Weight 228.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Hydroquinone Monosulfate