missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Sulfate, Fused, ACS Reagent, Reagents

Catalog No. p-200020965
Change view
Click to view available options
Quantity:
500 g
3 kg
Catalog No. Quantity
01337794 3 kg
01337793 500 g
2 options

Catalog No. 01337794

Supplier: Labchem Inc C12902003A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Hydrogen Sulfate, Fused, ACS Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
Synonym Potassium Hydrogen Sulfate
IUPAC Name dipotassium (sulfonatooxy)sulfonate
SMILES [K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1
Chemical Name or Material Potassium Hydrogen Sulfate, Fused, ACS Reagent