missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen sulfate, fused, ACS, mixture of potassium pyrosulfate, K{2}S{2}O{7}, and potassium bisulfate, KHSO{4}

Catalog No. p-4824892
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1380022 100 g
AA1380036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1380022 Supplier Thermo Scientific Chemicals Supplier No. 01380022
Only null left

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It is also used as a disintegrating agent in analytical chemistry. Further, it is used to make fertilizers and in the preparation of potassium persulfate. In addition to this, it is used as a catalyst in organic preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00066385

Specifications

Melting Point 214°C
Color White
Physical Form Fused Powder
Quantity 100 g
Solubility Information Soluble in water,isopropyl alcohol and acetone.
Formula Weight 136.17
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White fused powder
Chemical Name or Material Potassium hydrogen sulfate, fused
Show More Show Less
Hazard Category H314-H331
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8, Packing Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only