missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Phthalate 99.8+%, TCI America™

Catalog No. P0309500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0309500G 500 g
P030925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0309500G Supplier TCI America Supplier No. P0309500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 877-24-7
Color White
pH 4.0 (0.05M H2O soln. 25°C)
Physical Form Crystalline
Quantity 500 g
Synonym Monopotassium Phthalate, Phthalic Acid Monopotassium Salt
Molecular Weight (g/mol) 204.222
Percent Purity ≥99.8% (T)