missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Melting Point | 295.0°C to 300.0°C |
| Color | White |
| pH | 3.8 to 4 (5% soln. at 20°C) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.99% |
| Packaging | Plastic bottle |
| Molecular Formula | C8H5KO4 |
| Linear Formula | 2-(HO2C)C6H4CO2K |
| Quantity | 100 g |
| Beilstein | 09, 791 |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. NFPA Health: |
RUO – Research Use Only