missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phosphate, dried, 98+%

CAS: 7758-11-4 | HK2O4P | 174.17 g/mol

Supplier:  Thermo Scientific Chemicals A113210B

 View more versions of this product

Catalog No. AAA113210B

Only null left
Explore available promotions

Description

Description

Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen phosphate is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Hygroscopic chemical and incompatible with strong oxidizing agents and acids. Keep the material away from heat, sparks and flame and sources of ignition.
Specifications

Specifications

Potassium hydrogen phosphate
11-4-7758
HK2O4P
14,7658
Soluble in water. Slightly soluble in alcohol.
OP(=O)([O-])[O-].[K+].[K+]
174.17
CHEBI:32031
≥98%
Dried
1000 g
MFCD00011383
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium;hydrogen phosphate
24450
174.18
Hygroscopic

Chemical Identifiers

11-4-7758
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
dipotassium;hydrogen phosphate
HK2O4P
MFCD00011383
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
CHEBI:32031
OP(=O)([O-])[O-].[K+].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 231-834-5

RTECSNumber : TC5580000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only