missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phosphate, ACS, 98.0% min

Catalog No. p-4782080
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1159330 250 g
AA11593A1 1 kg
AA11593A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1159330 Supplier Thermo Scientific Chemicals Supplier No. 01159330
Only null left

CAS: 7758-11-4 | HK2O4P | 174.17 g/mol

It is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.0% min.
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 174.18
Grade ACS
Sensitivity Hygroscopic
Appearance White powder
Chemical Name or Material Potassium hydrogen phosphate
Hazard Category H319: Causes serious eye irritation.
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.
EINECSNumber 231-834-5
RTECSNumber TC5580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only