missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen carbonate, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 292.0°C |
| Quantity | 25 kg |
| Linear Formula | KHCO3 |
| Merck Index | 15,773 |
| Solubility Information | Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution |
| IUPAC Name | potassium hydrogen carbonate |
| Formula Weight | 100.12 |
| Grade | Extra Pure |
| Packaging | Fibre drum |
| Chemical Name or Material | Potassium hydrogen carbonate |
Safety and Handling
| EINECSNumber | 206-059- |
RUO – Research Use Only