missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen carbonate, MilliporeSigma™
Specifications
Specifications
| Color | White |
| pH | 8 to 8.6 (50 g/L at 20°C) |
| Physical Form | Solid |
| Quantity | 500 g |
| Solubility Information | Water: 333g/L at 20°C soluble |
| Formula Weight | 100.12 |
| Percent Purity | ≥99.5% |
| Grade | ACS |
| Odor | Odorless |
| Packaging | Plastic Bottle |
| Show More |
Safety and Handling
| EINECSNumber | 206-059-0 |
| RTECSNumber | FG1840000 |
| Recommended Storage | Tightly closed. Dry. |