missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen carbonate, ACS reagent
CAS: 298-14-6 | CHKO3 | 100.11 g/mol
Supplier: Thermo Scientific Chemicals 446300025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium hydrogen carbonate | |
| 298-14-6 | |
| CHKO3 | |
| MFCD00011402 | |
| potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| potassium;hydrogen carbonate | |
| 516893 | |
| 100.12 | |
| Plastic bottle |
| 292.0°C | |
| 2.5 kg | |
| KHCO3 | |
| 15,773 | |
| Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution | |
| [K+].OC([O-])=O | |
| 100.11 | |
| CHEBI:81862 | |
| ACS Reagent |
Chemical Identifiers
| 298-14-6 | |
| 100.11 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| 516893 | |
| [K+].OC([O-])=O |
| CHKO3 | |
| MFCD00011402 | |
| potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
| CHEBI:81862 |
Safety and Handling
EINECSNumber : 206-059-
RUO – Research Use Only