missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis)

Catalog No. AA4019536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4019536 500 g
AA4019522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4019536 Supplier Thermo Scientific Chemicals Supplier No. 04019536
Only null left

Maximum level of impurities: Insoluble matter 0.01%, Cl 0.001%, PO4 5ppm, SO4 0.003%, NH4 5ppm, Ca 0.002%, Mg 0.001%, Heavy Metals (as Pb) 5ppm, Fe 5ppm, Na 0.03% | CAS: 298-14-6 | CHKO3 | 100.11 g/mol

Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O

Specifications

Color White
Physical Form Granules
Quantity 500 g
Assay Percent Range 99.7-100.5%
Solubility Information Slightly soluble in water. Practically insoluble in alcoholSoluble in water. Insoluble in alcohol.
IUPAC Name potassium;hydrogen carbonate
Formula Weight 100.12
Grade ACS
Odor Odorless
Appearance White crystalline granules
Density 2.17 g/cm3 at 20°C
Chemical Name or Material Potassium hydrogen carbonate, Dried basis
Show More Show Less
EINECSNumber 206-059-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only