missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen carbonate, 99.5+%, for analysis

CAS: 298-14-6 | CHKO3 | 100.11 g/mol

$206.72 - $206.72

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC217590010
SDS View Documents
Thermo Scientific Chemicals
217590010
1 kg Plastic bottle
Each for $206.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

298-14-6
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
516893
[K+].OC([O-])=O
CHKO3
MFCD00011402
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
CHEBI:81862

Specifications

292.0°C
White
1 kg
CHKO3
MFCD00011402
15, 7730
Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution
[K+].OC([O-])=O
100.11
CHEBI:81862
99.5%
Plastic bottle
298-14-6
Crystalline Powder
99.5%
KHCO3
13,253
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium hydrogen carbonate
516893
100.12
Analytical
Potassium hydrogen carbonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-059-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only