missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen carbonate, 99.5+%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 292.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 5 kg |
| Assay Percent Range | 99.5% |
| Linear Formula | KHCO3 |
| Fieser | 13,253 |
| Merck Index | 15, 7730 |
| Solubility Information | Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C),practocally insoluble in alcohol,soluble in potassium carbonate solution |
| IUPAC Name | potassium;hydrogen carbonate |
| Show More |
Safety and Handling
| EINECSNumber | 206-059- |
RUO – Research Use Only