missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen carbonate, 99.5+%, for analysis

Catalog No. AC217590010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217590010 1 kg Plastic bottle
AC217590051 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC217590010 Supplier Thermo Scientific Chemicals Supplier No. 217590010
Only null left

CAS: 298-14-6 | CHKO3 | 100.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O

Specifications

Melting Point 292.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.5%
Linear Formula KHCO3
Fieser 13,253
Merck Index 15, 7730
Solubility Information Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution
IUPAC Name potassium hydrogen carbonate
Formula Weight 100.12
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium hydrogen carbonate
Show More Show Less
EINECSNumber 206-059-

RUO – Research Use Only