missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Potassium hydrogen carbonate, 99.5%, for biochemistry
CAS: 298-14-6 | CHKO3 | 100.11 g/mol
$121.02 - $121.02
Chemical Identifiers
| CAS | 298-14-6 |
|---|---|
| Molecular Formula | CHKO3 |
| Molecular Weight (g/mol) | 100.11 |
| MDL Number | MFCD00011402 |
| InChI Key | TYJJADVDDVDEDZ-UHFFFAOYSA-M |
| Synonym | potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 |
| PubChem CID | 516893 |
| ChEBI | CHEBI:81862 |
| SMILES | [K+].OC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC448015000
|
Thermo Scientific Chemicals
448015000 |
500 g |
Each for $121.02
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 298-14-6 | |
| 100.11 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| 516893 | |
| [K+].OC([O-])=O |
| CHKO3 | |
| MFCD00011402 | |
| potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
| CHEBI:81862 |
Specifications
| 292.0°C | |
| 500 g | |
| KHCO3 | |
| 15773 | |
| Clear colorless | |
| [K+].OC([O-])=O | |
| 100.11 | |
| CHEBI:81862 | |
| 99.5% | |
| Plastic Bottle |
| 298-14-6 | |
| CHKO3 | |
| MFCD00011402 | |
| potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| potassium hydrogen carbonate | |
| 516893 | |
| 100.12 | |
| Biochemical | |
| Potassium hydrogen carbonate |
Safety and Handling
EINECSNumber : 206-059-0
RUO – Research Use Only