missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexahydroxoantimonate(V), Honeywell Fluka™

Catalog No. p-7072232
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
6001226 100 g
6001227 500 g
2 options

Catalog No. 6001226

Supplier: Honeywell Chemicals 60500100G

Only null left

for the precipitation of sodium,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12208-13-8
MDL Number MFCD00011415

Specifications

Quantity 100 g
Linear Formula KSb(OH)6
UN Number UN1549
Formula Weight 262.9g/mol
Percent Purity ≥99%
Chemical Name or Material Potassium hexahydroxoantimonate(V)
Hazard Statement P273