missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorosilicate, 99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111019474
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111019474 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019474 Supplier Merck Emd Millipore Supplier No. 603221KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity reagent for laboratory and chemical applications.

Chemical Identifiers

CAS 16871-90-2
Molecular Formula F6K2Si
Molecular Weight (g/mol) 220.27
MDL Number MFCD00040666
InChI Key UUHPPUWEDCVPCO-UHFFFAOYSA-N
IUPAC Name dipotassium hexafluorosilanediuide
SMILES [K+].[K+].F[Si--](F)(F)(F)(F)F

Specifications

Physical Form Solid
Quantity 1 kg
Linear Formula K2SiF6
Formula Weight 220.27
Percent Purity ≥99.0% (T)
Packaging Poly Bottle
Density 2.665 g/mL (at 25°C)
Chemical Name or Material Potassium Hexafluorosilicate
RTECSNumber VV8400000