missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexafluorophosphate 95.0+%, TCI America™

Catalog No. p-7134907
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
P102325G 25 g
P1023100G 100 g
P1023500G 500 g
3 options

Catalog No. P102325G

Supplier: TCI America P102325G

Only null left

Chemical Identifiers

CAS 17084-13-8
Molecular Formula F6KP
Molecular Weight (g/mol) 184.062
MDL Number MFCD00011412
InChI Key YZDGRYDIGCWVND-UHFFFAOYSA-N
Synonym potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
PubChem CID 23688904
IUPAC Name potassium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[K+]

Specifications

Melting Point 575°C
Color White
pH 3.0 to 7.0 (50g/L H2O soln.)
Physical Form Crystalline Powder
Quantity 25 g
UN Number 3260
Formula Weight 184.06
Percent Purity ≥95.0% (T)
Chemical Name or Material Potassium Hexafluorophosphate
EINECSNumber (1)-0323
TSCA Yes