missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexafluorophosphate, Approx. 98%, Spectrum™ Chemical

Catalog No. p-7166120
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
140 lb.
2.5 kg
25 g
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Catalog No. Quantity Packaging
18612650 25 g Amber Glass Bottle
18612651 100 g Amber Glass Bottle
18612652 500 g Amber Glass Bottle
18612654 140 lb. Fiber Drum
18612653 2.5 kg Amber Glass Bottle
5 options

Catalog No. 18612650

Supplier: Spectrum Chemical Mfg Cor P180725GM

Only null left

Chemical Identifiers

CAS 17084-13-8

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity ∼98%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.