missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexafluorophosphate, Approx. 98%, Spectrum™ Chemical

Catalog No. 18612650
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
140 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612650 25 g Amber Glass Bottle
18612651 100 g Amber Glass Bottle
18612652 500 g Amber Glass Bottle
18612654 140 lb. Fiber Drum
18612653 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612650 Supplier Spectrum Chemical Mfg Cor Supplier No. P180725GM

Chemical Identifiers

CAS 17084-13-8

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity ∼98%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.