missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorophosphate, 99% min

CAS: 17084-13-8 | F6KP | 184.062 g/mol

Supplier:  Thermo Scientific Chemicals 04003030

 View more versions of this product

Catalog No. AA4003030

Only null left
Explore available promotions

Description

Description

Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium hexafluorophosphate
17084-13-8
Crystalline
99% min.
MFCD00011412
Soluble in water.
F[P-](F)(F)(F)(F)F.[K+]
184.062
184.07
Hygroscopic
2.55
575°C
White
250 g
F6KP
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium;hexafluorophosphate
23688904
Odorless
White Powder

Chemical Identifiers

17084-13-8
184.062
YZDGRYDIGCWVND-UHFFFAOYSA-N
23688904
F[P-](F)(F)(F)(F)F.[K+]
F6KP
MFCD00011412
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
potassium;hexafluorophosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c

H302+H312+H332-H314

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 241-143-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only