missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorophosphate, 99% min

Catalog No. AA4003022
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4003022 100 g
AA4003030 250 g
AA4003036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4003022 Supplier Thermo Scientific Chemicals Supplier No. 04003022

CAS: 17084-13-8 | F6KP | 184.062 g/mol

Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17084-13-8
Molecular Formula F6KP
Molecular Weight (g/mol) 184.062
MDL Number MFCD00011412
InChI Key YZDGRYDIGCWVND-UHFFFAOYSA-N
Synonym potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
PubChem CID 23688904
IUPAC Name potassium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[K+]

Specifications

Melting Point 575°C
Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99% min.
Solubility Information Soluble in water.
Formula Weight 184.07
Odor Odorless
Sensitivity Hygroscopic
Appearance White Powder
Density 2.55 g/cm3 at 20°C
Chemical Name or Material Potassium hexafluorophosphate
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P310-P312-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 241-143-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only