missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min

CAS: 339268-21-2 | C6K4N6Ru | 413.57 g/mol

$97.81 - $2794.48

Chemical Identifiers

CAS 339268-21-2
Molecular Formula C6K4N6Ru
Molecular Weight (g/mol) 413.57
MDL Number MFCD00168064
InChI Key FULSJSKUKUZHLZ-UHFFFAOYSA-N
Synonym potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate
PubChem CID 71311369
IUPAC Name tetrapotassium;ruthenium(2+);hexacyanide;hydrate
SMILES [K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11876SS
View Documents
Thermo Scientific Chemicals
011876SS
0.2 g
Each for $97.81
Only null left
 
AA1187604
View Documents
Thermo Scientific Chemicals
01187604
2 g
Each for $640.81
Only null left
 
AA1187609
View Documents
Thermo Scientific Chemicals
01187609
10 g
Each for $2,794.48
Only null left
 
Description

Description

Potassium hexacyanoruthenate(II) hydrate used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

339268-21-2
413.57
FULSJSKUKUZHLZ-UHFFFAOYSA-N
71311369
[K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
C6K4N6Ru
MFCD00168064
potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate
tetrapotassium;ruthenium(2+);hexacyanide;hydrate

Specifications

339268-21-2
Crystalline
C6K4N6Ru
MFCD00168064
potassium hexacyanoruthenate ii hydrate, tetrapotassium hexacyanorutheniumtetrauide hydrate
FULSJSKUKUZHLZ-UHFFFAOYSA-N
tetrapotassium;ruthenium(2+);hexacyanide;hydrate
71311369
Ru 23.0% min.
White
0.2 g
K4Ru(CN)6·xH2O
UN1588
Soluble in water.
[K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
413.57
413.57 (Anhydrous)
Potassium hexacyanoruthenate(II) hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.

EINECSNumber : 239-097-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only