missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(Iii), Honeywell Fluka™

Catalog No. p-7072012
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
6 x 1 kg
Catalog No. Quantity
6002332 250 g
6002333 500 g
6002331 1 kg
60045930 6 x 1 kg
4 options

Catalog No. 6002332

Supplier: Honeywell Chemicals 31253250G

Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Quantity 250 g
Linear Formula K3Fe(CN)6
IUPAC Name tripotassium;iron(3+);hexacyanide
Formula Weight 329.24g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Hexacyanoferrate(Iii), Reag. Ph. Eur. reag. ISO
EINECSNumber 237-323-3
RTECSNumber LJ8225000