missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hexacyanoferrate(III), For Analysis, EMSURE™, MilliporeSigma™

$382.14 - $382.14
Chemical Identifiers
CAS | 13746-66-2 |
---|---|
Molecular Formula | C6FeK3N6 |
Molecular Weight (g/mol) | 329.25 |
MDL Number | MFCD00011392 |
InChI Key | QMTKJUMXUDIUAQ-UHFFFAOYSA-N |
Synonym | potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559 |
PubChem CID | 26250 |
IUPAC Name | tripotassium hexacyanoirontris(ylium) |
SMILES | [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
M1049730250
|
Technidata
1049730250 |
250 g | Plastic Bottle |
Each for $382.14
|
|
|||||
Chemical Identifiers
13746-66-2 | |
329.25 | |
QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
26250 | |
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
C6FeK3N6 | |
MFCD00011392 | |
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559 | |
tripotassium hexacyanoirontris(ylium) |
Specifications
Red | |
6 (50g/L, H2O, 20°C) | |
250 g | |
MFCD00011392 | |
464g/L | |
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N | |
329.25 | |
329.25 | |
ACS | |
1.85Â g/3 (20Â °C) |
13746-66-2 | |
Solid | |
C6FeK3N6 | |
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559 | |
QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
tripotassium hexacyanoirontris(ylium) | |
26250 | |
>99.0% | |
Plastic Bottle |
Safety and Handling
EINECSNumber : 237-323-3