missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(III), For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1049730250
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049730250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1049730250 Supplier Technidata Supplier No. 1.04973.0250

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Red
pH 6 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 250 g
Solubility Information 464g/L
Formula Weight 329.25
Percent Purity >99.0%
Grade ACS
Packaging Plastic Bottle
Density 1.85 g/3 (20 °C)
EINECSNumber 237-323-3