Learn More
Potassium hexacyanocobaltate(III), 95%, Thermo Scientific Chemicals
$48.05 - $182.19
Chemical Identifiers
CAS | 13963-58-1 |
---|---|
Molecular Formula | C6CoK3N6 |
Molecular Weight (g/mol) | 332.34 |
MDL Number | MFCD00011391 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC203920050
|
Thermo Scientific Chemicals
203920050 |
5 g | Glass bottle |
Each for $48.05
|
|
||||
AC203921000
|
Thermo Scientific Chemicals
203921000 |
100 g | Glass bottle |
Each for $182.19
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13963-58-1 | |
C6CoK3N6 | |
332.34 | |
MFCD00011391 |
Specifications
Brown to Yellow | |
Crystalline Solid | |
95% | |
K3[Co(CN)6] | |
15, 7758 | |
332.34 | |
95% | |
1.87 | |
Potassium hexacyanocobaltate(III), 95% |
13963-58-1 | |
5 g | |
C6CoK3N6 | |
MFCD00011391 | |
Solubility in water: freely soluble. Other solubilities: insoluble in alcohol | |
332.34 | |
Glass bottle | |
1.8700g/mL |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Contact with acids liberates very toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 237-742-1
RTECSNumber : GF9470000
TSCA : TSCA
RUO – Research Use Only