missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexachloroiridate(IV), Ir 39% min
Description
Potassium hexachloroiridate(IV) is used to prepare platinum nanoparticles (PNPs) by reduction with trisodium citrate and sodium dodecyl sulfate as stabilizer reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Physical Form | Crystalline |
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 483.12 |
| Concentration or Composition (by Analyte or Components) | Ir 39% min. |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Potassium hexachloroiridate(IV) |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 240-976-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only