missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium heptafluorotantalate(V), 99.7%
Description
It is an important raw material. Used in chemical research. Use as primary and secondary intermediate(Pharmaceutical Intermediates). Used in the production and preparation of tantalum powder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 250 g |
| Density | 4.560 g/mL |
| UN Number | UN3288 |
| Solubility Information | Decomposes in water; Slightly soluble in HF |
| Formula Weight | 392.14 |
| Percent Purity | 99.7% |
| Chemical Name or Material | Potassium heptafluorotantalate(V) |
Safety and Handling
| Hazard Category | H301-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 240-986-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only