missing translation for 'onlineSavingsMsg'
Learn More

Potassium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. P24101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24101G 1 g
P24105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24101G Supplier TCI America Supplier No. P24101G

Chemical Identifiers

CAS 860653-59-4
Molecular Formula CF4KNO4S2
Molecular Weight (g/mol) 269.23
MDL Number MFCD28334505
InChI Key IMBHGOLFXVHBMC-UHFFFAOYSA-N
PubChem CID 45378297
IUPAC Name potassium (fluorosulfonyl)(trifluoromethanesulfonyl)azanide
SMILES [K+].FC(F)(F)S(=O)(=O)[N-]S(F)(=O)=O

Specifications

Melting Point 101°C
Color White
Quantity 1 g
UN Number 1759
Formula Weight 269.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (Fluorosulfonyl)(trifluoromethanesulfonyl)imide