missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluorosulfate, 99.5%

Catalog No. AA4032218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4032218 50 g
AA4032209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4032218 Supplier Thermo Scientific Chemicals Supplier No. 04032218

CAS: 13455-22-6 | FKO3S | 138.15 g/mol

Use as primary and secondary intermediate. Finds application in chemical research..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13455-22-6
Molecular Formula FKO3S
Molecular Weight (g/mol) 138.15
MDL Number MFCD00216618
InChI Key VXJAYNWISQFORV-UHFFFAOYSA-M
Synonym potassium fluorosulfate, potassium sulfurofluoridate, potassium fluorosulphate, fluorosulfonyloxypotassium, acmc-1c0t1, potassium fluorosulfonyl oxidanide, potassium ion o-fluorosulfonyl oxidanidolate, fluorosulfuric acid,potassium salt 8ci,9ci
PubChem CID 23678652
IUPAC Name potassium sulfurofluoridate
SMILES [K+].[O-]S(F)(=O)=O

Specifications

Melting Point 350°C (decomposition)
Color White
Physical Form Lump/Powder
Quantity 50 g
Linear Formula KSO3F
UN Number UN3260
Solubility Information Hydrolysis.
Formula Weight 138.16
Percent Purity 99.5%
Sensitivity Moisture sensitive
Chemical Name or Material Potassium fluorosulfate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only