missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, For Dissolved Oxygen, Certified, 40% (w/v), LabChem™
Specifications
Specifications
| Color | Clear, colorless |
| Format | Liquid |
| Molecular Formula | FK |
| MDL Number | MFCD00011398 |
| InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
| Packaging | Poly Bottle |
| SMILES | [F-].[K+] |
| IUPAC Name | potassium fluoride |
| Molecular Weight (g/mol) | 58.10 |
| Quantity | 4 L |
| Show More |