missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, For Dissolved Oxygen, Certified, 40% (w/v), LabChem™

Catalog No. LC191001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC191001 500 mL
LC191002 1 L
LC191004 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC191001 Supplier LabChem Supplier No. LC191001
Only null left

Specifications

Color Clear, colorless
Format Liquid
Molecular Formula FK
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Packaging Poly Bottle
SMILES [F-].[K+]
IUPAC Name potassium fluoride
Molecular Weight (g/mol) 58.10
Quantity 500 mL
Storage Requirements Room Temperature
Type Potassium Fluoride
Show More Show Less