missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride Dihydrate, Reagent Grade, Reagents
Potassium Fluoride Dihydrate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$863.60 - $3022.73
Chemical Identifiers
| CAS | 13455-21-5 |
|---|---|
| Molecular Formula | FH4KO2 |
| Molecular Weight (g/mol) | 94.13 |
| MDL Number | MFCD00149929 |
| InChI Key | MZFXHSQFQLKKFD-UHFFFAOYSA-M |
| IUPAC Name | potassium dihydrate fluoride |
| SMILES | O.O.[F-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-845
|
Labchem Inc
C128400025A5 |
2.5 kg |
Each for $3,022.73
|
|
|||||
|
01-337-844
|
Labchem Inc
C1284000500A5 |
500 g |
Each for $863.60
|
|
|||||
Chemical Identifiers
| 13455-21-5 | |
| 94.13 | |
| MZFXHSQFQLKKFD-UHFFFAOYSA-M | |
| O.O.[F-].[K+] |
| FH4KO2 | |
| MFCD00149929 | |
| potassium dihydrate fluoride |
Specifications
| 156°C | |
| 13455-21-5 | |
| 2.5 kg | |
| MFCD00149929 | |
| O.O.[F-].[K+] | |
| 94.13 | |
| Poly Bottle | |
| Potassium Fluoride Dihydrate, Reagent Grade |
| Colorless/White | |
| Solid | |
| FH4KO2 | |
| MZFXHSQFQLKKFD-UHFFFAOYSA-M | |
| potassium dihydrate fluoride | |
| Reagent | |
| 2.45 |