missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals

$62.32 - $414.85

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1155514
View Documents
Thermo Scientific Chemicals
01155514
25 g
Each for $62.32
Only null left
 
AA1155530
View Documents
Thermo Scientific Chemicals
01155530
250 g
Each for $129.40
Only null left
 
AA11555A1
View Documents
Thermo Scientific Chemicals
011555A1
1 kg
Each for $414.85
Only null left
 
Description

Description

Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13455-21-5
94.13
MZFXHSQFQLKKFD-UHFFFAOYSA-M
23708904
FH4KO2
MFCD00149929
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
O.O.[F-].[K+]

Specifications

41°C
Crystalline
FH4KO2
UN1812
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium dihydrate fluoride
23708904
≥98.5%
1.352
2.5 g/mL
13455-21-5
25 g
MFCD00149929
14,7632
Freely soluble in waterSoluble in water; Insoluble in ethanol.
O.O.[F-].[K+]
94.13
94.13 (58.10 Anhydrous)
Odorless
Hygroscopic
Potassium fluoride dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c

H301+H311+H331-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID

EINECSNumber : 232-151-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only