missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals

Catalog No. AA11555A1
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11555A1 1 kg
AA1155514 25 g
AA1155530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11555A1 Supplier Thermo Scientific Chemicals Supplier No. 011555A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]

Specifications

Melting Point 41°C
Physical Form Crystalline
Quantity 1 kg
UN Number UN1812
Merck Index 14,7632
Solubility Information Freely soluble in waterSoluble in water; Insoluble in ethanol.
IUPAC Name potassium;fluoride;dihydrate
Formula Weight 94.13 (58.10 Anhydrous)
Percent Purity ≥98.5%
Odor Odorless
Refractive Index 1.352
Sensitivity Hygroscopic
Density 2.5 g/mL
Chemical Name or Material Potassium fluoride dihydrate
Show More Show Less
Hazard Category H301+H311+H331-H318
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID
EINECSNumber 232-151-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only