missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, Purified (Potassium Hexacyanoferrate (III)), Reagents

Catalog No. 01337110
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337110 100 g
01337109 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337110 Supplier Labchem Inc Supplier No. C2283000100C2
Only null left

Potassium Ferricyanide, Purified (Potassium Hexacyanoferrate (III)), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym Potassium Hexacyanoferrate III, Red Prussiate, Potassium Ferricyanate
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Ruby Red
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.89
Chemical Name or Material Potassium Ferricyanide, Purified (Potassium Hexacyanoferrate (III))