missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferricyanide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| Formula Weight | 329.26 |
| Percent Purity | ≥99.5% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.85kg |
| Chemical Name or Material | Potassium Ferricyanide |