missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III)), Reagents

Catalog No. 01337849
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337849 100 g
01337848 1 kg
01337847 25 kg
01337846 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337849 Supplier Labchem Inc Supplier No. C1283000100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III)), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym Potassium Hexacyanoferrate III, Red Prussiate, Potassium Ferricyanate
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 300°C
Color Orange/Red
Physical Form Solid
Quantity 100 g
Percent Purity 100%
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.89
Chemical Name or Material Potassium Ferricyanide, ACS Reagent (Potassium Hexacyanoferrate (III))