missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. LC190401
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC190401 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC190401 Supplier LabChem Supplier No. LC190401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Orange
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula K3Fe(CN)6
Solubility Information Soluble in water
Formula Weight 329.26
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.85g/cm3
Decomposition Information Potassium oxide; Carbon monoxide; Carbon dioxide
Density 1.85g/cm3
Chemical Name or Material Potassium Ferricyanide
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature