missing translation for 'onlineSavingsMsg'
Learn More

Potassium ferricyanide, 98%, pure

Catalog No. p-43102
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196781000 100 g Glass bottle
AC196785000 500 g Glass bottle
AC196780010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC196781000 Supplier Thermo Scientific Chemicals Supplier No. 196781000
Only null left

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Orange to Red
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula K3Fe(CN)6
Fieser 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255
Merck Index 15, 7751
Solubility Information Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name tripotassium;iron(3+);hexacyanide
Formula Weight 329.26
Percent Purity 98%
Grade Pure
Packaging Glass bottle
Density 1.85
Chemical Name or Material Potassium ferricyanide, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.
Precautionary Statement GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 237-323-3
RTECSNumber LJ8225000
TSCA TSCA

RUO – Research Use Only