missing translation for 'onlineSavingsMsg'
Learn More

Potassium ethyltrifluoroborate, 97%

Catalog No. AC444880050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444880050 5 g
AC444880010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444880050 Supplier Thermo Scientific Chemicals Supplier No. 444880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 44248-07-9 | C2H5BF3K | 135.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 44248-07-9
Molecular Formula C2H5BF3K
Molecular Weight (g/mol) 135.97
MDL Number MFCD04112713
InChI Key GIIPADVFWNGSKY-UHFFFAOYSA-N
Synonym potassium ethyltrifluoroborate, potassium ethyltrifluoroboranuide, potassium ethytrifluoroborate, amtb738, ethyltrifluoroborate potassium salt, potassium ethyl trifluoro borate 1-
PubChem CID 23668491
IUPAC Name potassium;ethyl(trifluoro)boranuide
SMILES [K+].CC[B-](F)(F)F

Specifications

Melting Point 80.0°C to 84.0°C
Assay Percent Range 97%
Quantity 5 g
Formula Weight 135.97
Percent Purity 97%
Chemical Name or Material Potassium ethyltrifluoroborate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only