missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Disulfate, For Analysis, EMSURE™, MilliporeSigma™
Specifications
Specifications
| Melting Point | 325°C |
| Color | White |
| pH | 0.8 (50g/L, H2O, 20°C) |
| Physical Form | Solid |
| Quantity | 5 kg |
| Formula Weight | 254.33 |
| Percent Purity | 97.5 to 100.0% |
| Grade | ACS |
| Packaging | Plastic Bottle |
Safety and Handling
| EINECSNumber | 232-216-8 |