missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Potassium Dichromate Solution Standard, Set of 2

Catalog No. 142982071
Change view
Click to view available options
Quantity:
Set of 2
Set of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
142982071 Set of 2
142982072 Set of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 142982071 Supplier Thermo Scientific™ Supplier No. 9423UV95100E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Dichromate Solution Standards are NIST-traceable for qualifying instrument performance.

Thermo Scientific™ Potassium Dichromate Solution Standards are NIST-traceable for qualifying instrument performance. Qualify your spectrophotometer's performance with NIST-traceable Thermo Scientific Potassium Dichromate Solution Standards for photometric accuracy.

The Potassium Dichromate Solution Standard is available in a set of two or six standards. Both standard sets are:
  • Supplied in 0.001M perchloric acid in permanently sealed quartz cells
  • Compatible with any cell holder that accommodates 10mm pathlength rectangular cells
  • Traceable to NIST™ standards and supplied with a calibration certificate
  • Calibrated for use with instruments with a SBW of ≤1.6nm, including the Thermo Scientific Evolution 60, Evolution 300 and Evolution 600

Set of Two Standards

  • Intended for testing photometric accuracy at 430nm as required by the European Pharmacopoeia Section 2.2.25, revised 2005
  • Includes a 600mg/L standard and corresponding blank

Set of Six Standards

  • Designed to check linearity of the instrument in the UV region
  • Calibrated at 235, 257, 313 and 350nm
  • Includes standards at 0, 20, 40, 60, 80 and 100mg/L concentrations

Specifications

Calibration NIST Traceable
Wavelength Range 235 to 350 nm
For Use With (Equipment) Instruments with a SBW of ≤1.6nm; Evolution 60; Evolution 300; Evolution 600
Includes 0 and 60mg/L standards
Quantity Set of 2