missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, 0.250 N (N/4), 0.04167 M (M/24), Ricca Chemical
APHA/ASTM/EPA for COD Testing
Supplier: Ricca Chemical Company RI60705
Specifications
| Orange | |
| Liquid | |
| 20L |
| 0.250Normal,0.04167Molar | |
| Cubitainer | |
| Potassium Dichromate |