missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium (cyanomethyl)trifluoroborate, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 888711-58-8 |
|---|---|
| Molecular Formula | C2H2BF3KN |
| Molecular Weight (g/mol) | 146.95 |
| InChI Key | IJENQMHWPRCCBA-UHFFFAOYSA-N |
| Synonym | potassium cyanomethyl trifluoroborate, c2h2bf3n.k, potassium cyanomethyl trifluoroboranuide, potassium cyanomethyl trifluoro borate 1- |
| PubChem CID | 23666193 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC444860010
|
Thermo Scientific Chemicals
444860010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC444860050
|
Thermo Scientific Chemicals
444860050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 888711-58-8 | |
| 146.95 | |
| potassium cyanomethyl trifluoroborate, c2h2bf3n.k, potassium cyanomethyl trifluoroboranuide, potassium cyanomethyl trifluoro borate 1- |
| C2H2BF3KN | |
| IJENQMHWPRCCBA-UHFFFAOYSA-N | |
| 23666193 |
Specifications
| 888711-58-8 | |
| Glass Bottle | |
| 1g | |
| IJENQMHWPRCCBA-UHFFFAOYSA-N | |
| 23666193 | |
| 95% |
| 95% | |
| C2H2BF3KN | |
| potassium cyanomethyl trifluoroborate, c2h2bf3n.k, potassium cyanomethyl trifluoroboranuide, potassium cyanomethyl trifluoro borate 1- | |
| 146.95 | |
| 146.95 | |
| Potassium (cyanomethyl)trifluoroborate |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only