missing translation for 'onlineSavingsMsg'
Learn More

Potassium (cyanomethyl)trifluoroborate, 95%

Catalog No. AC444860010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444860010 1 g
AC444860050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444860010 Supplier Thermo Scientific Chemicals Supplier No. 444860010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 888711-58-8 | C2H2BF3KN | 146.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 888711-58-8
Molecular Formula C2H2BF3KN
Molecular Weight (g/mol) 146.95
InChI Key IJENQMHWPRCCBA-UHFFFAOYSA-N
Synonym potassium cyanomethyl trifluoroborate, c2h2bf3n.k, potassium cyanomethyl trifluoroboranuide, potassium cyanomethyl trifluoro borate 1-
PubChem CID 23666193

Specifications

Melting Point 151°C to 156°C
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 146.95
Percent Purity 95%
Chemical Name or Material Potassium (cyanomethyl)trifluoroborate
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only