missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Cyanide, Technical, 96%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 500 g |
| Percent Purity | 96% |
| Grade | Technical |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food, drug or pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.